Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATAAAAATGATGACAGTCGGCC[A/G]TGGCTCACCTGTTCCTGCCAGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ITFG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITFG2 - integrin alpha FG-GAP repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018463.3 | 305 | Silent Mutation | CCA,CCG | P,P 55 | NP_060933.3 | |
XM_005253707.4 | 305 | Silent Mutation | CCA,CCG | P,P 55 | XP_005253764.1 | |
XM_005253709.4 | 305 | Intron | XP_005253766.1 | |||
XM_017019641.1 | 305 | Intron | XP_016875130.1 |
NRIP2 - nuclear receptor interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |