Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTAATCATGTTTCTGACAATC[A/G]TAGTGTGTGGAATGGTTGCTGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605659 | ||||||||||||||||||||
Literature Links: |
CLEC2D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC2D - C-type lectin domain family 2 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004419.4 | 169 | Missense Mutation | ATA,GTA | I,V 48 | NP_001004419.1 | |
NM_001197317.2 | 169 | Intron | NP_001184246.1 | |||
NM_001197318.2 | 169 | Missense Mutation | ATA,GTA | I,V 48 | NP_001184247.1 | |
NM_001197319.2 | 169 | Intron | NP_001184248.1 | |||
NM_013269.5 | 169 | Missense Mutation | ATA,GTA | I,V 48 | NP_037401.1 |