Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTGGTGGACGTGGGGGGCGAGTA[C/T]GACCCTCGGAGACACCGATATGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605378 MIM: 611366 MIM: 604143 MIM: 604899 | ||||||||||||||||||||
Literature Links: |
AAAS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AAAS - aladin WD repeat nucleoporin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C12orf10 - chromosome 12 open reading frame 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021640.3 | 346 | Silent Mutation | TAC,TAT | Y,Y 98 | NP_067653.3 |
ESPL1 - extra spindle pole bodies like 1, separase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN5 - prefoldin subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002624.3 | 346 | Intron | NP_002615.2 | |||
NM_145897.2 | 346 | Intron | NP_665904.1 |