Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCCAAGCGTTATAATCGCAAAC[A/G]TGAACTTTCCTACCCCAAAAATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615998 | ||||||||||||||||||||
Literature Links: |
RNF10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF10 - ring finger protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014868.4 | 703 | Missense Mutation | CAT,CGT | H,R 67 | NP_055683.3 | |
XM_005254013.2 | 703 | Missense Mutation | CAT,CGT | H,R 17 | XP_005254070.1 | |
XM_006719717.2 | 703 | Missense Mutation | CAT,CGT | H,R 67 | XP_006719780.1 | |
XM_017020282.1 | 703 | Missense Mutation | CAT,CGT | H,R 17 | XP_016875771.1 | |
XM_017020283.1 | 703 | Missense Mutation | CAT,CGT | H,R 67 | XP_016875772.1 |