Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGGGCCCTGTATTGGGCCTG[C/T]GTCCACAATGATCCCACCCAGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD33 - ankyrin repeat domain 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130015.1 | 407 | UTR 5 | NP_001123487.1 | |||
NM_001304459.1 | 407 | UTR 5 | NP_001291388.1 | |||
NM_001304460.1 | 407 | UTR 5 | NP_001291389.1 | |||
NM_182608.3 | 407 | Silent Mutation | TGC,TGT | C,C 103 | NP_872414.3 | |
XM_017019258.1 | 407 | Silent Mutation | TGC,TGT | C,C 112 | XP_016874747.1 |