Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGAGTTCCTGGAGCTGAGCACA[C/G]AACAGTCAGTTTTTCACATAGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615053 MIM: 610681 | ||||||||||||||||||||
Literature Links: |
ASB8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASB8 - ankyrin repeat and SOCS box containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319296.1 | 951 | Missense Mutation | CTG,GTG | L,V 243 | NP_001306225.1 | |
NM_001319297.1 | 951 | Missense Mutation | CTG,GTG | L,V 243 | NP_001306226.1 | |
NM_001319298.1 | 951 | Missense Mutation | CTG,GTG | L,V 182 | NP_001306227.1 | |
NM_001319299.1 | 951 | Missense Mutation | CTG,GTG | L,V 182 | NP_001306228.1 | |
NM_001319300.1 | 951 | UTR 3 | NP_001306229.1 | |||
NM_001319301.1 | 951 | UTR 3 | NP_001306230.1 | |||
NM_024095.4 | 951 | Missense Mutation | CTG,GTG | L,V 243 | NP_077000.1 |
PFKM - phosphofructokinase, muscle | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |