Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTTGGCGGTACTTATGTTTTCC[C/T]GACTCAAAGTTGGCCAGCATCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601958 MIM: 612172 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNB3 - calcium voltage-gated channel auxiliary subunit beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX23 - DEAD-box helicase 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004818.2 | Intron | NP_004809.2 |