Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTCATTCCTCTATTTACAGAC[C/G]TGCCGCAGACAATTCTGCTAGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616988 MIM: 616989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLLU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLLU1 - chronic lymphocytic leukemia up-regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025233.2 | 763 | Missense Mutation | CCT,CGT | P,R 14 | NP_001020404.1 |
CLLU1OS - chronic lymphocytic leukemia up-regulated 1 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025232.1 | 763 | Intron | NP_001020403.1 | |||
XM_017019673.1 | 763 | Intron | XP_016875162.1 |