Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCACTGGTCCTTGCCCCTAGTTG[G/T]TGGTGGGAGGCCTGCTGTCTCCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612819 | ||||||||||||||||||||
Literature Links: |
DDX51 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX51 - DEAD-box helicase 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOC4L - nucleolar complex associated 4 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024078.2 | 524 | Missense Mutation | GTG,TTG | V,L 153 | NP_076983.1 | |
XM_017019960.1 | 524 | Missense Mutation | GTG,TTG | V,L 153 | XP_016875449.1 |