Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTATTAGCACGTGGCATACAC[C/T]TACCAACATGTTTTGAACATCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607363 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KNTC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KNTC1 - kinetochore associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014708.4 | Intron | NP_055523.1 | ||||
XM_006719706.2 | Intron | XP_006719769.1 | ||||
XM_011539027.2 | Intron | XP_011537329.1 | ||||
XM_011539029.2 | Intron | XP_011537331.1 | ||||
XM_011539030.1 | Intron | XP_011537332.1 | ||||
XM_017020254.1 | Intron | XP_016875743.1 | ||||
XM_017020255.1 | Intron | XP_016875744.1 | ||||
XM_017020256.1 | Intron | XP_016875745.1 | ||||
XM_017020257.1 | Intron | XP_016875746.1 |
RSRC2 - arginine and serine rich coiled-coil 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |