Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAATGTTTAGATAGTTACCTGTT[C/G]ATAAACTGACAAGTATCCCCACTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608044 | ||||||||||||||||||||
Literature Links: |
SLC5A8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC5A8 - solute carrier family 5 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145913.3 | 2015 | Missense Mutation | TCA,TGA | S,* 542 | NP_666018.3 | |
XM_017018910.1 | 2015 | Intron | XP_016874399.1 |