Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCTTTTTGAAGGTTTAATTCC[A/G]CATAGGTTATTTCCTGTTCAGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 161555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLRC1 - killer cell lectin like receptor C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304448.1 | 579 | Missense Mutation | GCG,GTG | A,V 41 | NP_001291377.1 | |
NM_002259.4 | 579 | Missense Mutation | GCG,GTG | A,V 41 | NP_002250.1 | |
NM_007328.3 | 579 | Missense Mutation | GCG,GTG | A,V 41 | NP_015567.1 | |
NM_213657.2 | 579 | Missense Mutation | GCG,GTG | A,V 41 | NP_998822.1 | |
NM_213658.2 | 579 | Missense Mutation | GCG,GTG | A,V 41 | NP_998823.1 |