Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCAAGGGGTTGGGCAGCAGCTC[C/T]AATCTTCCCAAATGAGTATAGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606677 | ||||||||||||||||||||
Literature Links: |
CLEC4C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC4C - C-type lectin domain family 4 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130441.2 | 369 | Nonsense Mutation | TAG,TGG | *,W 80 | NP_569708.1 | |
NM_203503.1 | 369 | Nonsense Mutation | TAG,TGG | *,W 49 | NP_987099.1 | |
XM_017018940.1 | 369 | Nonsense Mutation | TAG,TGG | *,W 80 | XP_016874429.1 |