Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGCCCAGCTCCTGCACTGCTG[C/T]AGTTGTATTATCCAGCTTTCTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605070 | ||||||||||||||||||||
Literature Links: |
EEA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEA1 - early endosome antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003566.3 | 4238 | Missense Mutation | ACA,GCA | T,A 1325 | NP_003557.2 | |
XM_011538814.2 | 4238 | Missense Mutation | ACA,GCA | T,A 1367 | XP_011537116.1 | |
XM_011538815.2 | 4238 | Missense Mutation | ACA,GCA | T,A 1279 | XP_011537117.1 | |
XM_017020018.1 | 4238 | Missense Mutation | ACA,GCA | T,A 1331 | XP_016875507.1 |
PLEKHG7 - pleckstrin homology and RhoGEF domain containing G7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |