Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAAAGCTGGCTGCGCCACCTC[C/T]GCACACAGTGGTGGAGTTGCCAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123940 MIM: 611160 | ||||||||||||||||||||
Literature Links: |
KRT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT4 - keratin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT79 - keratin 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175834.2 | 1465 | Missense Mutation | AGA,GGA | R,G 478 | NP_787028.1 |