Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACGGAGCCCAGTTGGCCATTCC[G/T]CCTCTCGTCGTATGTGGCAAAGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 154050 MIM: 603887 | ||||||||||||||||||||
Literature Links: |
MIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIP - major intrinsic protein of lens fiber | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012064.3 | 236 | Silent Mutation | NP_036196.1 | |||
XM_011538354.1 | 236 | Missense Mutation | XP_011536656.1 | |||
XM_017019306.1 | 236 | Missense Mutation | XP_016874795.1 |
TIMELESS - timeless circadian clock | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |