Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCATCTCCCATTTGGCGTCATGC[A/T]GCTCTGGGTCTGGTAACTCTTTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CLEC12B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC12B - C-type lectin domain family 12 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129998.2 | 350 | Silent Mutation | GCA,GCT | A,A 42 | NP_001123470.1 | |
NM_001319241.1 | 350 | UTR 5 | NP_001306170.1 | |||
NM_001319242.1 | 350 | UTR 5 | NP_001306171.1 | |||
NM_205852.3 | 350 | Silent Mutation | GCA,GCT | A,A 42 | NP_995324.2 | |
XM_011520658.2 | 350 | Silent Mutation | GCA,GCT | A,A 33 | XP_011518960.1 | |
XM_011520663.2 | 350 | Silent Mutation | GCA,GCT | A,A 42 | XP_011518965.1 | |
XM_017019295.1 | 350 | UTR 5 | XP_016874784.1 |
LOC102724020 - uncharacterized LOC102724020 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |