Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGAGCCTCCAGCAGAGATGGCT[A/G]AGGTCCAGTAGCTGATTTTGTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIGNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FIGNL2 - fidgetin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013690.4 | 3764 | UTR 3 | NP_001013712.4 | |||
XM_005268870.3 | 3764 | UTR 3 | XP_005268927.1 | |||
XM_017019298.1 | 3764 | UTR 3 | XP_016874787.1 | |||
XM_017019299.1 | 3764 | UTR 3 | XP_016874788.1 |
SCN8A - sodium voltage-gated channel alpha subunit 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |