Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGATGAACAGAATCTGCCGGAT[G/T]GTGCTGGCCACTTGCTTGGGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST11 - carbohydrate (chondroitin 4) sulfotransferase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173982.1 | 549 | Missense Mutation | ATG,ATT | M,I 17 | NP_001167453.1 | |
NM_018413.5 | 549 | Missense Mutation | ATG,ATT | M,I 17 | NP_060883.1 | |
XM_017019369.1 | 549 | Intron | XP_016874858.1 |