Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCATCCATGAGCGCTACCACT[C/T]AGTCCTGCAGCCACTGGACATAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124050 | ||||||||||||||||||||
Literature Links: |
DAO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAO - D-amino acid oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001917.4 | 317 | Missense Mutation | TCA,TTA | S,L 25 | NP_001908.3 | |
XM_005268692.3 | 317 | Missense Mutation | TCA,TTA | S,L 25 | XP_005268749.1 | |
XM_011538004.2 | 317 | Missense Mutation | TCA,TTA | S,L 25 | XP_011536306.1 | |
XM_011538005.2 | 317 | Missense Mutation | TCA,TTA | S,L 25 | XP_011536307.1 |