Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTAACCATCTTTGGATCTTTGT[A/G]TCCTAAGATGTTCAAAAAACTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600372 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEDD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEDD1 - neural precursor cell expressed, developmentally down-regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135175.1 | Intron | NP_001128647.1 | ||||
NM_001135176.1 | Intron | NP_001128648.1 | ||||
NM_001135177.1 | Intron | NP_001128649.1 | ||||
NM_152905.3 | Intron | NP_690869.1 | ||||
XM_005268644.2 | Intron | XP_005268701.1 | ||||
XM_006719237.3 | Intron | XP_006719300.1 | ||||
XM_011537903.2 | Intron | XP_011536205.1 | ||||
XM_011537904.2 | Intron | XP_011536206.1 | ||||
XM_011537905.2 | Intron | XP_011536207.1 | ||||
XM_011537906.2 | Intron | XP_011536208.1 | ||||
XM_017018801.1 | Intron | XP_016874290.1 | ||||
XM_017018802.1 | Intron | XP_016874291.1 |