Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCAAAATAAACCTGTGTCCTG[A/C]CCTTTCTTCATTAATTGTAGCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRIG3 - leucine rich repeats and immunoglobulin like domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136051.2 | Intron | NP_001129523.1 | ||||
NM_153377.4 | Intron | NP_700356.2 | ||||
XM_011537893.2 | Intron | XP_011536195.1 | ||||
XM_017018790.1 | Intron | XP_016874279.1 |