Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACTTGGCCTTGGAATTCCAGCT[A/G]CTGCTGCTTTTTAGGAAGCCCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603281 | ||||||||||||||||||||
Literature Links: |
C12orf43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf43 - chromosome 12 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OASL - 2'-5'-oligoadenylate synthetase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261825.1 | 1694 | Nonsense Mutation | CAG,TAG | Q,* 345 | NP_001248754.1 | |
NM_003733.3 | 1694 | Nonsense Mutation | CAG,TAG | Q,* 475 | NP_003724.1 | |
NM_198213.2 | 1694 | UTR 3 | NP_937856.1 | |||
XM_017020140.1 | 1694 | Intron | XP_016875629.1 | |||
XM_017020141.1 | 1694 | Intron | XP_016875630.1 |