Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTACATGTTCATCTGGAAGATGGC[A/G]CCCGTCGTCTGGCACAGGAGGTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606926 | ||||||||||||||||||||
Literature Links: |
LPAR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPAR5 - lysophosphatidic acid receptor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142961.1 | 575 | Silent Mutation | NP_001136433.1 | |||
NM_020400.5 | 575 | Silent Mutation | NP_065133.1 |