Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126337 MIM: 156560 MIM: 614489 | ||||||||||||||||||||
Literature Links: |
DDIT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDIT3 - DNA damage inducible transcript 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195053.1 | 448 | Silent Mutation | ACC,ACG | T,T 60 | NP_001181982.1 | |
NM_001195054.1 | 448 | Silent Mutation | ACC,ACG | T,T 60 | NP_001181983.1 | |
NM_001195055.1 | 448 | Silent Mutation | ACC,ACG | T,T 60 | NP_001181984.1 | |
NM_001195056.1 | 448 | Silent Mutation | ACC,ACG | T,T 60 | NP_001181985.1 | |
NM_001195057.1 | 448 | Silent Mutation | ACC,ACG | T,T 37 | NP_001181986.1 | |
NM_004083.5 | 448 | Silent Mutation | ACC,ACG | T,T 37 | NP_004074.2 |
MARS - methionyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004990.3 | 448 | Intron | NP_004981.2 | |||
XM_006719398.3 | 448 | Intron | XP_006719461.1 |
MBD6 - methyl-CpG binding domain protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR616 - microRNA 616 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6758 - microRNA 6758 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |