Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCCCTTCTTGGAGCGGGCACCC[C/T]GGCCGAAGGCGCCCTGCTGCAGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611665 | ||||||||||||||||||||
Literature Links: |
CFAP73 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFAP73 - cilia and flagella associated protein 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144872.1 | 1158 | UTR 3 | NP_001138344.1 | |||
XM_011538327.2 | 1158 | Intron | XP_011536629.1 |
DDX54 - DEAD-box helicase 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111322.1 | 1158 | Missense Mutation | CAG,CGG | Q,R 869 | NP_001104792.1 | |
NM_024072.3 | 1158 | Missense Mutation | CAG,CGG | Q,R 868 | NP_076977.3 |
MIR7106 - microRNA 7106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |