Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTAGCACAGGCTGGCACAGCC[A/G]CTGGGCAGGGGGCTGGCACCCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 125855 MIM: 155550 | ||||||||||||||||||||
Literature Links: |
DGKA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGKA - diacylglycerol kinase alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMEL - premelanosome protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200053.1 | 1540 | Missense Mutation | CGG,TGG | R,W 453 | NP_001186982.1 | |
NM_001200054.1 | 1540 | Missense Mutation | CGG,TGG | R,W 539 | NP_001186983.1 | |
NM_001320121.1 | 1540 | Missense Mutation | CGG,TGG | R,W 497 | NP_001307050.1 | |
NM_001320122.1 | 1540 | Missense Mutation | CGG,TGG | R,W 497 | NP_001307051.1 | |
NM_006928.4 | 1540 | Missense Mutation | CGG,TGG | R,W 539 | NP_008859.1 | |
XM_006719569.1 | 1540 | Missense Mutation | CGG,TGG | R,W 539 | XP_006719632.1 | |
XM_011538685.1 | 1540 | Missense Mutation | CGG,TGG | R,W 539 | XP_011536987.1 |