Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATAATAAATGCCTAAGTTACAGG[A/G]ACAGAACCAATCTTTCATTAATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBX5 - chromobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127321.1 | 5637 | UTR 3 | NP_001120793.1 | |||
NM_001127322.1 | 5637 | UTR 3 | NP_001120794.1 | |||
NM_012117.2 | 5637 | UTR 3 | NP_036249.1 |
MIR3198-2 - microRNA 3198-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |