Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGAAGACAAGGTAGCCCCCACC[A/G]TGGAGACCTCTGACCCTTATGCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606239 MIM: 179514 MIM: 606887 | ||||||||||||||||||||
Literature Links: |
IKZF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IKZF4 - IKAROS family zinc finger 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105369781 - uncharacterized LOC105369781 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUOX - sulfite oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000456.2 | 1380 | Missense Mutation | ATG,GTG | M,V 170 | NP_000447.2 | |
NM_001032386.1 | 1380 | Missense Mutation | ATG,GTG | M,V 170 | NP_001027558.1 | |
NM_001032387.1 | 1380 | Missense Mutation | ATG,GTG | M,V 170 | NP_001027559.1 | |
XM_005269112.1 | 1380 | Missense Mutation | ATG,GTG | M,V 177 | XP_005269169.1 | |
XM_017019905.1 | 1380 | Missense Mutation | ATG,GTG | M,V 177 | XP_016875394.1 | |
XM_017019906.1 | 1380 | Missense Mutation | ATG,GTG | M,V 177 | XP_016875395.1 | |
XM_017019907.1 | 1380 | Missense Mutation | ATG,GTG | M,V 170 | XP_016875396.1 | |
XM_017019908.1 | 1380 | Missense Mutation | ATG,GTG | M,V 170 | XP_016875397.1 |