Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCTATCCCCCAGGGCCACTTAC[C/T]GGGTTTTCCCATGGCCTTCATGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602113 | ||||||||||||||||||||
Literature Links: |
KMT2D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KMT2D - lysine methyltransferase 2D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHEBL1 - Ras homolog enriched in brain like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303126.1 | 640 | Missense Mutation | CAG,CGG | Q,R 109 | NP_001290055.1 | |
NM_144593.2 | 640 | Missense Mutation | CAG,CGG | Q,R 111 | NP_653194.1 | |
XM_017018794.1 | 640 | Missense Mutation | CAG,CGG | Q,R 111 | XP_016874283.1 | |
XM_017018795.1 | 640 | Missense Mutation | CAG,CGG | Q,R 109 | XP_016874284.1 |