Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGCTGTGAGACCACCCCCTCAA[A/G]GGGCAGCACCCCCACCCTCACACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX54 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX54 - DEAD-box helicase 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IQCD - IQ motif containing D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RITA1 - RBPJ interacting and tubulin associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286215.1 | 921 | Missense Mutation | AAG,AGG | K,R 110 | NP_001273144.1 | |
NM_032848.2 | 921 | Missense Mutation | AAG,AGG | K,R 86 | NP_116237.1 | |
XM_005253972.2 | 921 | Missense Mutation | AAG,AGG | K,R 86 | XP_005254029.1 |