Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGGAAGCTTTGCTGGAATGGC[A/C]TGCACATAACCCAACCACATAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609841 MIM: 609666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC8B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC8B1 - solute carrier family 8 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719607.2 | Intron | XP_006719670.1 | ||||
XM_011538749.2 | Intron | XP_011537051.1 | ||||
XM_011538750.2 | Intron | XP_011537052.1 | ||||
XM_011538752.2 | Intron | XP_011537054.1 | ||||
XM_017019978.1 | Intron | XP_016875467.1 |
TPCN1 - two pore segment channel 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |