Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTGCACACGGTGCTTCTGCA[C/T]CGCAGCACAGGCAAGTTCCACTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615089 | ||||||||||||||||||||
Literature Links: |
ATG101 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG101 - autophagy related 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098673.1 | 397 | Silent Mutation | CAC,CAT | H,H 31 | NP_001092143.1 | |
NM_021934.4 | 397 | Silent Mutation | CAC,CAT | H,H 31 | NP_068753.2 |
OR7E47P - olfactory receptor family 7 subfamily E member 47 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |