Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCAGTGGAATAAAGGTAGTAT[A/G]TGTTTTCCGCTGCTTCCGTGCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605861 | ||||||||||||||||||||
Literature Links: |
CNPY2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNPY2 - canopy FGF signaling regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAN2 - PAN2 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127460.2 | 3246 | Missense Mutation | CAT,TAT | H,Y 957 | NP_001120932.1 | |
NM_001166279.1 | 3246 | Missense Mutation | CAT,TAT | H,Y 956 | NP_001159751.1 | |
NM_014871.4 | 3246 | Missense Mutation | CAT,TAT | H,Y 953 | NP_055686.3 |