Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAACCTCGCCAAGTCTCAGACG[C/T]TGCTGGGGAAGGAAATCAAAATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ULK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ULK1 - unc-51 like autophagy activating kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003565.2 | 443 | Silent Mutation | CTG,TTG | L,L 59 | NP_003556.1 | |
XM_011538798.2 | 443 | Silent Mutation | CTG,TTG | L,L 59 | XP_011537100.1 | |
XM_011538799.2 | 443 | Silent Mutation | CTG,TTG | L,L 59 | XP_011537101.1 |