Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGGCAACCAGACCTCAGACTTT[A/G]AGAACACTCTGGGGTCAGCATTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DNAJC22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC22 - DnaJ heat shock protein family (Hsp40) member C22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304944.1 | 1136 | Missense Mutation | AAG,GAG | K,E 132 | NP_001291873.1 | |
NM_024902.3 | 1136 | Missense Mutation | AAG,GAG | K,E 132 | NP_079178.2 | |
XM_005269155.2 | 1136 | Missense Mutation | AAG,GAG | K,E 132 | XP_005269212.1 | |
XM_005269156.2 | 1136 | Missense Mutation | AAG,GAG | K,E 132 | XP_005269213.1 | |
XM_005269157.2 | 1136 | Missense Mutation | AAG,GAG | K,E 132 | XP_005269214.1 |