Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGTGGGCGAGCTCGGCCGCCAG[A/G]TGGAGTGGGGAGCGCAGCTGTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605476 MIM: 609677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGAP2 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122772.2 | 2410 | Silent Mutation | CAC,CAT | H,H 1096 | NP_001116244.1 | |
NM_014770.3 | 2410 | Silent Mutation | CAC,CAT | H,H 740 | NP_055585.1 | |
XM_005268625.3 | 2410 | Silent Mutation | CAC,CAT | H,H 1076 | XP_005268682.1 | |
XM_005268626.2 | 2410 | Silent Mutation | CAC,CAT | H,H 760 | XP_005268683.1 | |
XM_017018772.1 | 2410 | Silent Mutation | CAC,CAT | H,H 1096 | XP_016874261.1 |
AGAP2-AS1 - AGAP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OS9 - OS9, endoplasmic reticulum lectin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |