Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGATGAGCTTACTACAGGCAGC[A/G]CAGTTTCCCGTTACACCAAAGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180386 | ||||||||||||||||||||
Literature Links: |
LMO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMO3 - LIM domain only 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001395.2 | 585 | Silent Mutation | TGC,TGT | C,C 77 | NP_001001395.1 | |
NM_001243609.1 | 585 | Silent Mutation | TGC,TGT | C,C 77 | NP_001230538.1 | |
NM_001243610.1 | 585 | Silent Mutation | TGC,TGT | C,C 77 | NP_001230539.1 | |
NM_001243611.1 | 585 | Silent Mutation | TGC,TGT | C,C 88 | NP_001230540.1 | |
NM_001243612.1 | 585 | Silent Mutation | TGC,TGT | C,C 95 | NP_001230541.1 | |
NM_001243613.1 | 585 | Silent Mutation | TGC,TGT | C,C 99 | NP_001230542.1 | |
NM_018640.4 | 585 | Silent Mutation | TGC,TGT | C,C 77 | NP_061110.2 | |
XM_006719110.3 | 585 | Silent Mutation | TGC,TGT | C,C 77 | XP_006719173.1 | |
XM_006719111.2 | 585 | Silent Mutation | TGC,TGT | C,C 77 | XP_006719174.1 | |
XM_011520762.1 | 585 | Silent Mutation | TGC,TGT | C,C 77 | XP_011519064.1 | |
XM_011520763.1 | 585 | Silent Mutation | TGC,TGT | C,C 77 | XP_011519065.1 | |
XM_017019643.1 | 585 | Silent Mutation | TGC,TGT | C,C 57 | XP_016875132.1 |