Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGGAGAGATCCACAACTGGAC[C/T]GAGCTGCTTGACCTCTTCAACCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605307 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR182 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR182 - G protein-coupled receptor 182 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007264.3 | 860 | Silent Mutation | ACC,ACT | T,T 30 | NP_009195.1 | |
XM_011537824.2 | 860 | Silent Mutation | ACC,ACT | T,T 30 | XP_011536126.1 | |
XM_011537825.2 | 860 | Silent Mutation | ACC,ACT | T,T 30 | XP_011536127.1 | |
XM_011537827.2 | 860 | Silent Mutation | ACC,ACT | T,T 30 | XP_011536129.1 |
HBCBP - HBcAg-binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB39 - zinc finger and BTB domain containing 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |