Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGCCAAAGAGCAGCAGGCGGA[A/G]GCAGAGCTCCGGAAGCTAAGGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC63 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC63 - coiled-coil domain containing 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286243.1 | 221 | UTR 5 | NP_001273172.1 | |||
NM_001286244.1 | 221 | Intron | NP_001273173.1 | |||
NM_152591.2 | 221 | Silent Mutation | GAA,GAG | E,E 28 | NP_689804.1 | |
XM_006719262.1 | 221 | Silent Mutation | GAA,GAG | E,E 28 | XP_006719325.1 | |
XM_006719263.2 | 221 | Silent Mutation | GAA,GAG | E,E 28 | XP_006719326.1 | |
XM_011537999.1 | 221 | Silent Mutation | GAA,GAG | E,E 28 | XP_011536301.1 | |
XM_011538001.2 | 221 | Silent Mutation | GAA,GAG | E,E 28 | XP_011536303.1 |