Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGAAGTGTGGAGACGGAACGC[C/G]AGGGCGAAAGCCAAGGCCAATGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
H1FNT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
H1FNT - H1 histone family member N, testis specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181788.1 | 840 | Silent Mutation | GCC,GCG | A,A 176 | NP_861453.1 |
ZNF641 - zinc finger protein 641 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |