Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACTATTCTGTATACTTCTTGGG[A/C]AAAGTTGAAAAAGAAATCCTTTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616929 | ||||||||||||||||||||
Literature Links: |
MTERF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTERF2 - mitochondrial transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033050.2 | 1132 | Missense Mutation | TGC,TTC | C,F 259 | NP_001028222.1 | |
NM_025198.4 | 1132 | Missense Mutation | TGC,TTC | C,F 259 | NP_079474.2 | |
XM_011538767.2 | 1132 | Missense Mutation | TGC,TTC | C,F 259 | XP_011537069.1 | |
XM_017019985.1 | 1132 | Missense Mutation | TGC,TTC | C,F 259 | XP_016875474.1 | |
XM_017019986.1 | 1132 | Missense Mutation | TGC,TTC | C,F 259 | XP_016875475.1 | |
XM_017019987.1 | 1132 | Missense Mutation | TGC,TTC | C,F 259 | XP_016875476.1 |
TMEM263 - transmembrane protein 263 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |