Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCATCCGGCCGGGGGACTTCTC[C/T]CGCTCCAGGGGGCGGCCAGGAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605629 MIM: 602740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIT - citron rho-interacting serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206999.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2020 | NP_001193928.1 | |
NM_007174.2 | 6140 | Silent Mutation | CGA,CGG | R,R 1978 | NP_009105.1 | |
XM_006719206.2 | 6140 | Silent Mutation | CGA,CGG | R,R 2005 | XP_006719269.1 | |
XM_011537783.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2035 | XP_011536085.1 | |
XM_011537784.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2034 | XP_011536086.1 | |
XM_011537785.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2020 | XP_011536087.1 | |
XM_011537787.1 | 6140 | Silent Mutation | CGA,CGG | R,R 1963 | XP_011536089.1 | |
XM_011537788.1 | 6140 | Silent Mutation | CGA,CGG | R,R 1962 | XP_011536090.1 | |
XM_011537789.1 | 6140 | Silent Mutation | CGA,CGG | R,R 1589 | XP_011536091.1 | |
XM_011537790.2 | 6140 | Silent Mutation | CGA,CGG | R,R 1589 | XP_011536092.1 | |
XM_011537791.2 | 6140 | Silent Mutation | CGA,CGG | R,R 1569 | XP_011536093.1 | |
XM_011537792.2 | 6140 | Silent Mutation | CGA,CGG | R,R 1496 | XP_011536094.1 | |
XM_017018735.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2019 | XP_016874224.1 | |
XM_017018736.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2019 | XP_016874225.1 | |
XM_017018737.1 | 6140 | Silent Mutation | CGA,CGG | R,R 2004 | XP_016874226.1 | |
XM_017018738.1 | 6140 | Silent Mutation | CGA,CGG | R,R 1527 | XP_016874227.1 | |
XM_017018739.1 | 6140 | Silent Mutation | CGA,CGG | R,R 1496 | XP_016874228.1 |
PRKAB1 - protein kinase AMP-activated non-catalytic subunit beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |