Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAAAGTGAAACCCCATCCCCCT[C/T]CTTCTGGTTTACCTCCTTGTCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607926 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLT8D2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLT8D2 - glycosyltransferase 8 domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HCFC2 - host cell factor C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013320.2 | 458 | Missense Mutation | CCT,TCT | P,S 119 | NP_037452.1 | |
XM_017019241.1 | 458 | Missense Mutation | CCT,TCT | P,S 119 | XP_016874730.1 | |
XM_017019242.1 | 458 | Missense Mutation | CCT,TCT | P,S 119 | XP_016874731.1 |