Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAATTGTTGAAGAGTTCAACTT[C/T]CCCAGAAGTTGTTCTAACGCTGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609539 | ||||||||||||||||||||
Literature Links: |
ARID2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARID2 - AT-rich interaction domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152641.2 | 234 | Silent Mutation | TTC,TTT | F,F 78 | NP_689854.2 | |
XM_006719272.3 | 234 | Silent Mutation | TTC,TTT | F,F 78 | XP_006719335.1 | |
XM_011538025.1 | 234 | Intron | XP_011536327.1 |
LINC00938 - long intergenic non-protein coding RNA 938 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |