Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGTACAATGGTGCTCCCTTTA[A/T]GATTTTTAAAAATTTTACTTACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608102 MIM: 605653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLN5 - ceroid-lipofuscinosis, neuronal 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXL3 - F-box and leucine rich repeat protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012158.2 | 2299 | UTR 3 | NP_036290.1 | |||
XM_005266336.1 | 2299 | UTR 3 | XP_005266393.1 | |||
XM_005266337.2 | 2299 | UTR 3 | XP_005266394.1 | |||
XM_017020538.1 | 2299 | Intron | XP_016876027.1 |