Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTGCATCAAATTCCCAAAACTA[C/T]GCCTTTGAAGTAAGTTGACATAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAP18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAP18 - Sin3A associated protein 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKA3 - spindle and kinetochore associated complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166017.1 | 2396 | UTR 3 | NP_001159489.1 | |||
NM_145061.5 | 2396 | UTR 3 | NP_659498.4 | |||
XM_005266288.3 | 2396 | UTR 3 | XP_005266345.1 | |||
XM_011534994.1 | 2396 | UTR 3 | XP_011533296.1 |