Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATCCTACCCTTGAATGACAAGG[A/G]GACACTAAGTCCCTTTCTGCCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604331 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INTS6 - integrator complex subunit 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5693 - microRNA 5693 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINE3 - serpin family E member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101320.1 | Intron | NP_001094790.1 | ||||
XM_017020707.1 | Intron | XP_016876196.1 |