Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGAAGTTATCAGGGAGTAACTG[C/T]CCATCTAAAAGAAACTGGTCATCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 133280 | ||||||||||||||||||||
Literature Links: |
ESD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESD - esterase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001984.1 | 943 | Silent Mutation | GGA,GGG | G,G 232 | NP_001975.1 | |
XM_005266278.2 | 943 | Silent Mutation | GGA,GGG | G,G 232 | XP_005266335.1 | |
XM_011534954.1 | 943 | Silent Mutation | GGA,GGG | G,G 232 | XP_011533256.1 | |
XM_017020414.1 | 943 | Silent Mutation | GGA,GGG | G,G 113 | XP_016875903.1 |