Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAAGCAATGCACTGCAGCAACC[C/T]CAAGAGTGGAGTTGTGCTGGCTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604036 | ||||||||||||||||||||
Literature Links: |
CCNA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCNA1 - cyclin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111045.1 | 155 | Missense Mutation | CCC,CTC | P,L 49 | NP_001104515.1 | |
NM_001111046.1 | 155 | Missense Mutation | CCC,CTC | P,L 6 | NP_001104516.1 | |
NM_001111047.1 | 155 | Missense Mutation | CCC,CTC | P,L 6 | NP_001104517.1 | |
NM_003914.3 | 155 | Missense Mutation | CCC,CTC | P,L 50 | NP_003905.1 | |
XM_011535294.2 | 155 | Missense Mutation | CCC,CTC | P,L 6 | XP_011533596.1 | |
XM_011535295.2 | 155 | Missense Mutation | CCC,CTC | P,L 6 | XP_011533597.1 | |
XM_011535296.2 | 155 | Missense Mutation | CCC,CTC | P,L 6 | XP_011533598.1 |